missing translation for 'onlineSavingsMsg'
Learn More

CLIP Mouse anti-Human, Unlabeled, Clone: CERCLIP, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 555980

Product Code. 15848568

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Reacts with the class II associated invariant chain peptides (CLIP). CLIP is the remaining segment of class II invariant chain (Ii) after proteolytic degradation which stays associated with HLA-DR and represents a final intermediate in the removal of invariant chain from class II molecules during antigen processing. The removal of CLIP and subsequent loading of the antigenic peptide is facilitated by the non-classical class II molecule HLA-DM. CLIP can be detected, in association with HLA-DR, on the surface of T2DR3, a mutant cell line lacking HLA-DM.

Host Species: Mouse
Clone: CerCLIP
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry, Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CLIP
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Flow Cytometry
CERCLIP
Unconjugated
Mouse
0.1 mg
Immunology
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.