Learn More
Clec9A (CD370) Rat anti-Human, PE, Clone: 3A4/Clec9A (also known as 20/05-3A4 or 3A4), BD
Rat Monoclonal Antibody
Brand: BD Biosciences 563488
Description
The 3A4/Clec9A monoclonal antibody specifically binds to human Clec9A (C-type lectin domain family member 9A). Clec9A is a type II transmembrane glycoprotein with a single extracellular C-type lectin domain. Clec9A is also known as CD370 and DNGR1 (Dendritic cell natural killer lectin group receptor 1). Clec9A is selectively expressed on CD141+ myeloid dendritic cells. Clec9A reportedly functions as an endocytic receptor for necrotic cells. It can mediate the cross-presentation of dead-cell associated antigens by dendritic cells in a Syk-dependent manner. The 3A4 antibody reportedly crossreacts with peripheral blood dendritic cells form Pigtail macaques ( Macaca nemestrina ).
Specifications
| Clec9A (CD370) | |
| Monoclonal | |
| 0.2mg/mL | |
| CD370; C-type lectin domain family 9 member A; CLEC9A; DNGR1 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Human, Monkey | |
| IgG2a κ |
| Flow Cytometry | |
| 3A4/Clec9A (also known as 20/05-3A4 or 3A4) | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human Clec9A Peptide | |
| 25 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.