missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLDND1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542670
This item is not returnable.
View return policy
Description
Peptide sequence: TLTEQFMEKF VDPGNHNSGI DLLRTYLWRC QFLLPFVSLG LMCFGALIGL Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
CLDND1 is widely distributed in the adult CNS with highest expression in the corpus callosum, caudate nucleus, cerebral cortex, medulla, putamen, spinal cord, substantia nigra and subthalamic nucleus. Weak expression has been detected in the adult heart. There are 2 named isoforms produced by alternative splicing.
Specifications
| CLDND1 | |
| Polyclonal | |
| Unconjugated | |
| CLDND1 | |
| 1110019C08Rik; AA407103; AI849195; AW489850; C3orf4; claudin 25; claudin domain containing 1; claudin domain containing 1 protein; claudin domain-containing protein 1; Claudin-25; Cldn25; CLDND1; D11Moh34; GENX-3745; HSPC174; Membrane protein GENX-3745; MGC111162; MGC3316; MGC9861; PSEC0054; UNQ2511/PRO6000 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56650 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NY35 | |
| CLDND1 | |
| Synthetic peptide directed towards the middle region of human CLDND1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction