missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin 12 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA525768
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine and mouse based on sequence homology.
The claudin superfamily consists of many structurally related proteins in humans. These proteins, which include claudin 1 through 18, are important structural and functional components of tight junctions in paracellular transport. Claudins are located in both epithelial and endothelial cells in all tight junction-bearing tissues. Claudins, which consist of four trans-membrane domains and two extracellular loops, make up tight junction strands.Specifications
Claudin 12 | |
Polyclonal | |
Unconjugated | |
CLDN12 | |
claudin-12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9069 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.43 mg/mL | |
PBS with 0.09% sodium azide | |
P56749 | |
CLDN12 | |
KLH conjugated synthetic peptide between 19-46 amino acids from the N-terminal region of human CLDN12 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |