missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLTCL1 Polyclonal Antibody

Description
In direct ELISAs, less than 1% cross-reactivity with recombinant human CHC17 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
Specifications
Specifications
| Antigen | CLTCL1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | CLTCL1 |
| Gene Accession No. | P53675 |
| Gene Alias | CHC22; clathrin heavy chain 2; clathrin heavy chain like 1; clathrin heavy chain on chromosome 22; clathrin, heavy chain-like 1; Clathrin, heavy polypeptide D; clathrin, heavy polypeptide-like 1; CLH22; CLH-22; CLTCL; CLTCL1; CLTD |
| Gene Symbols | CLTCL1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?