missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHRND Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546828
This item is not returnable.
View return policy
Description
Peptide sequence: LTLGLLAALA VCGSWGLNEE ERLIRHLFQE KGYNKELRPV AHKEESVDVA Sequence homology: Cow: 91%; Dog: 100%; Guinea Pig: 93%; Human: 100%; Pig: 100%; Rabbit: 86%; Rat: 85%; Zebrafish: 78%.
The acetylcholine receptor of muscle has 5 subunits of 4 different types: 2 alpha and 1 each of beta, gamma and delta subunits. After acetylcholine binding, the receptor undergoes an extensive conformation change that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
Specifications
| CHRND | |
| Polyclonal | |
| Unconjugated | |
| CHRND | |
| acetylcholine receptor; acetylcholine receptor delta subunit; acetylcholine receptor subunit delta; acetylcholine receptor, nicotinic, delta (muscle); AChR delta subunit; Achr-4; ACHRD; Acrd; cholinergic receptor; cholinergic receptor nicotinic delta subunit; cholinergic receptor, nicotinic delta; cholinergic receptor, nicotinic, delta (muscle); cholinergic receptor, nicotinic, delta polypeptide; CHRND; CMS2A; CMS3A; CMS3B; CMS3C; delta subunit of muscle acetylcholine receptor; FCCMS; L10076; nicotinic acetylcholine receptor delta polypeptide; SCCMS; sofa potato; sop; wu:fa01e01; zgc:110469 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1144 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q07001 | |
| CHRND | |
| Synthetic peptide directed towards the N-terminal of human CHRND (aa 6-55). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction