Learn More
Chondroitin Sulfate Mouse anti-Human, Unlabeled, Clone: 9.2.27, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554275
Description
Tumor cells display a variety of antigens which have been defined by antibodies. Antibodies that specifically react with tumor-associated antigens are useful in understanding the biology of particular tumor types. Monoclonal antibody 9.2.27, produced against a human melanoma cell extract, reacts with a chondroitin sulfate proteoglycan. Proteoglycans including chondroitin sulfate, keratan sulfate, dermatan sulfate and heparan sulfate are major components of the extracellular matrices of many animal cell types. Clone 9.2.27 has been shown to react with human melanoma cells, glioma cells, and proliferating brain endothelial cells. It did not react with fetal melanocytes, neuroblastoma LA-N-1 cells or a variety of carcinoma, lymphoid, and fibroblastoid cell lines. Additionally, neither white or gray matter from the medulla oblongata, cerebellum or spinal cord of a normal human adult reacted with 9.2.27. Clone 9.2.27 has also been used in functional studies to investigate the role of chondroitin sulfate proteoglycans in human tumor cell systems. It has been shown to block melanoma cell spreading, exhibit antiproliferative effects on glioma cells in vitro, and suppress glioma tumor growth in athymic nude mice models.
A soluble extract derived from M14 human melanoma cells bound to lens culinaris lectin-Sepharose and depleted of fibronectin was used as immunogen.
Host Species: Mouse
Clone: 9.2.27
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Human Melanoma Cells
Electron Microscopy, Flow Cytometry, Immunofluorescence, Immunoprecipitation
Specifications
| Chondroitin Sulfate | |
| Monoclonal | |
| 0.5mg/mL | |
| CSPG4; NG2; Chondroitin sulfate proteoglycan 4; MCSP; MCSPG; MELCSPG; MSK16 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| 9.2.27 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human Melanoma Cells | |
| 0.1 mg | |
| Cell Biology | |
| Human | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.