missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHK2 Mouse anti-Human, Clone: 20A8, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517218
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
A suggested positive control for this product is HeLa cells.
Both Chk1 and Chk2 are critically important checkpoint kinases. Chk1 is essential for normal development and cell growth and Chk1 deficiency in mouse or in embryonic stem (ES) cells is lethal. Loss of Chk2 is tolerated and has been found to be associated with a subset of Li-Fraumeni cases. Li-Fraumeni syndrome increases greatly the susceptibility to cancer, with particularly high occurrences of breast cancer, brain tumors, acute leukemia, soft tissue sarcomas, bone sarcomas, and adrenal cortical carcinoma. Chk2 regulates cell cycle checkpoints and apoptosis in response to DNA damage, particularly to DNA double-strand breaks. The cell either stops the cell cycle and therefore its proliferation until the damage is repaired, or it destructs itself (apoptosis). The ataxia telangiectasia mutated (ATM) and ATR (ATM and Rad3-related) protein kinases exert cell cycle delay, in part, by phosphorylating Chk1, Chk2, and p53. Phosphorylation on Chk1 and p53 requires ATR, whereas phosphorylation on Chk2 requires ATM. Screening of novel substrates of Chk1 and Ckh2 followed by imunoprecipitation kinase assay and site-directed mutagenesis analysis suggests that HDAC5 and PGC-1 are specific targets for Chk1 and/or Chk2 at least in vitro.
Spezifikation
| Chk2 | |
| Monoclonal | |
| 1 mg/mL | |
| HEPES with 0.01% BSA, 0.15M NaCl, 50% glycerol and 0.03% sodium azide | |
| O96017 | |
| CHEK2 | |
| Recombit human protein purified from E.coli(His-Chk2) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunoprecipitation, Western Blot | |
| 20A8 | |
| Unconjugated | |
| CHEK2 | |
| CDS1; CHK2; HuCds1; LFS2; PP1425; RAD53; hCds1; CHK2 checkpoint homolog; cds1 homolog; checkpoint-like protein CHK2; protein kinase CHK2; serine/threonine-protein kinase Chk2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 11200 | |
| -20°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur