missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CGGBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530276
This item is not returnable.
View return policy
Description
Recommended positive controls: Jurkat. Predicted reactivity: Mouse (98%), Rat (100%), Rhesus Monkey (99%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CGGBP1 influences expression of the FMR1 gene (MIM 309550), which is associated with the fragile X mental retardation syndrome (MIM 300624), by specifically interacting with the 5-prime (CGG)n-3-prime repeat in its 5-prime UTR.
Specifications
| CGGBP1 | |
| Polyclonal | |
| Unconjugated | |
| Cggbp1 | |
| 20 kDa CGG-binding protein; AA960172; AL023996; AW045841; CGG triplet repeat binding protein 1; CGG triplet repeat-binding protein 1; CGG-binding protein 1; CGGBP; Cggbp1; p20-CGG binding protein; p20-CGGBP; p20-CGGBP DNA-binding protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8545 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.78 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9UFW8 | |
| Cggbp1 | |
| Recombinant protein encompassing a sequence within the center region of human CGGBP1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction