missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ceruloplasmin Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA127046
This item is not returnable.
View return policy
Description
Store as a concentrated solution. Centrifuge briefly prior to opening vial.
Ceruloplasmin encodes a protein that is a metalloprotein that binds most of the copper in plasma and is involved in the peroxidation of Fe(II)transferrin to Fe(III) transferrin. Mutations in this gene cause aceruloplasminemia, which results in iron accumulation and tissue damage, and is associated with diabetes and neurologic abnormalities.
Specifications
| Ceruloplasmin | |
| Polyclonal | |
| Unconjugated | |
| CP | |
| bilitranslocase; CERP; Ceruloplasmin; ceruloplasmin (ferroxidase); CP; CP-2; D3Ertd555e; ferroxidase | |
| Goat | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation | |
| Conc. Not Determined | |
| PBS with no preservative; pH 7.3 | |
| P00450 | |
| CP | |
| Full length protein (Human). | |
| RUO | |
| 1356 | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction