Learn More
cerberus 1, cysteine knot superfamily, homolog (Xenopus laevis), Mouse, Clone: 3E11, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CER1.
Brand: Abnova H00009350-M04.100ug
Description
This gene encodes a cytokine member of the cysteine knot superfamily, characterized by nine conserved cysteines and a cysteine knot region. The cerberus-related cytokines, together with Dan and DRM/Gremlin, represent a group of bone morphogenetic protein (BMP) antagonists that can bind directly to BMPs and inhibit their activity. [provided by RefSeq
Sequence: HWETCRTVPFSQTITHEGCEKVVVQNNLCFGKCGSVHFPGAAQHSHTSCSHCLPAKFTTMHLPLNCTELSSVIKVVMLVEECQCKVKTEHEDGHILHAGSQDSFIPGVSSpecifications
| cerberus 1, cysteine knot superfamily, homolog (Xenopus laevis) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_005454 | |
| CER1 | |
| CER1 (NP_005445.1, 158 a.a. ∼ 266 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Stem Cell Biology | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunoprecipitation, Western Blot | |
| 3E11 | |
| Mouse monoclonal antibody raised against a full length recombinant CER1. | |
| CER1 | |
| DAND4/MGC119894/MGC119895/MGC96951 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 9350 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.