missing translation for 'onlineSavingsMsg'
Learn More
Learn More
centrosomal protein 55kDa, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human CEP55 protein.
Brand: Abnova H00055165-D01P.100ug
This item is not returnable.
View return policy
Description
Sequence: MSSRSTKDLIKSKWGSKPSNSKSETTLEKLKGEIAHLKTSVDEITSGKGKLTDKERHRLLEKIRVLEAEKEKNAYQLTEKDKEIQRLRDQLKARYSTTALLEQLEETTREGERREQVLKALSEEKDVLKQQLSAATSRIAELESKTNTLRLSQTVAPNCFNSSINNIHEMEIQLKDALEKNQQWLVYDQQREVYVKGLLAKIFELEKKTETAAHSLPQQTKKPESEGYLQEEKQKCYNDLLASAKKDLEVERQTITQLSFELSEFRRKYEETQKEVHNLNQLLYSQRRADVQHLEDDRHKTEKIQKLREENDIARGKLEEEKKRSEELLSQVQFLYTSLLKQQEEQTRVALLEQQMQACTLDFENEKLDRQHVQHQLLVILKELRKARNQITQLESLKQLHEFAITEPLVTFQGETENREKVAASPKSPTAALNESLVECPKCNIQYPATEHRDLLVHVEYCSKSpecifications
| centrosomal protein 55kDa | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human CEP55 protein. | |
| CEP55 | |
| C10orf3/FLJ10540/URCC6 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55165 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_018131.3 | |
| CEP55 | |
| CEP55 (NP_060601.2, 1 a.a. ∼ 464 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction