missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM Pan Mouse anti-Human, Clone: TET2, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA117760
This item is not returnable.
View return policy
Description
This antibody recognizes native human CEACAM1, 5, 6, 8 protein expressed in transfected mammalian cells.
CEA-related cell adhesion molecules (CEACAM) belong to the carcinoembryonic antigen (CEA) family. The CEA family proteins belong to the immuno-globulin (Ig) superfamily and are composed of one Ig variable-like (IgV) and a varying number (0-6) of Ig constant-like (IgC) domains. CEACAM molecules are membrane-bound either via a transmembrane domain or a glycosyl phosphatidyl inositol (GPI) anchor. CEACAM molecules are differentially expressed in epithelial cells or in leucocytes. Over-expression of CEA/CEACAM5 in tumors of epithelial origin is the basis of its wide-spread use as a tumor marker. The function of CEACAM family members varies widely: they function as cell adhesion molecules, tumor suppressors, regulators of lymphocyte and dendritic cell activation, receptors of Neisseria species and other bacteria.
Specifications
| CEACAM Pan | |
| Monoclonal | |
| 2 mg/mL | |
| CEACAM1 | |
| pan CEACAM, Carcinoembryonic antigen-related cell adhesion molecule 1, BGP, BGP-1, CD66a, Biliary glycoprotein 1, Carcinoembryonic antigen-related cell adhesion molecule 5, CEA, CD66e, Carcinoembryonic antigen-related cell adhesion molecule 6, CD66c, NCA, Carcinoembryonic antigen-related cell adhesion molecule 8, CD67, Carcinoembryonic antigen CGM6, CGM6, CD66b | |
| Mouse | |
| Protein G | |
| RUO | |
| 1048, 1088, 4680, 634 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot | |
| TET2 | |
| Unconjugated | |
| P06731, P13688, P31997, P40199 | |
| CEACAM1, CEACAM5, CEACAM6, CEACAM8 | |
| extracted protein of CEACAM5 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction