missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDX2 Monoclonal Antibody (EPR2764Y), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA514494
This item is not returnable.
View return policy
Description
MA5-14494 targets CDX2 in WB, IHC (P), ICC/IF and FACS applications and shows reactivity with Human and Mouse samples. The MA5-14494 immunogen is a synthetic peptide corresponding to residues near the N-terminus of human CDX2.
The product of this gene is an epsilon subunit of coatomer protein complex. Coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles. It is required for budding from Golgi membranes, and is essential for the retrograde Golgi-to-ER transport of dilysine-tagged proteins. Coatomer complex consists of at least the alpha, beta, beta', gamma, delta, epsilon and zeta subunits. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| CDX2 | |
| Monoclonal | |
| Conc. Not Determined | |
| tissue culture supernatant with 0.09% sodium azide | |
| P43241, Q99626 | |
| CDX2 | |
| A synthetic peptide corresponding to residues near the N-terminus of human CDX2 | |
| RUO | |
| 1045, 12591 | |
| 4° C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| EPR2764Y | |
| Unconjugated | |
| CDX2 | |
| caudal type homeo box 2; caudal type homeo box transcription factor 2; caudal type homeobox 2; caudal type homeobox transcription factor 2; caudal-type homeobox protein 2; CDX2; Cdx-2; CDX2/AS; CDX3; CDX-3; HGNC:1806; homeobox protein CDX-2; homeobox protein miniCDX2; homeobox transcription factor | |
| Rabbit | |
| 1 mL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction