Learn More
CDX-2 Mouse anti-Human, PE, Clone: M39-71, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 563428
Description
The M39-711 monoclonal antibody reacts with caudal-type homeobox transcription factor 2 (CDX-2). In embryonic development, CDX-2 is first expressed in the outer layer of the morula and later in the trophoblast, but not in embryonic stem cells. Along with Oct3/4, it is involved in the segregation of the trophoectoderm lineage from the inner cell mass. In the adult, CDX-2 expression is limited to the mucosa of the small and large intestines, where it regulates the synthesis of several intestine-specific proteins. Loss of CDX-2 expression is observed in some colorectal carcinomas, while gain of CDX-2 expression has been observed in gastric tumors and acute myeloid leukemias.
Specifications
| CDX-2 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human CDX-2 Recombinant Protein | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| M39-71 | |
| CDX2, CDX3, CDX-3 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.