Learn More
CDw210 Rat anti-Human, Unlabeled, Clone: 3F9, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 556012
Description
Reaacts with 90-110kDa protein expressed on T cells, B cells, NK cells, and monocyte/macrophages. It belongs to the class II cytokine receptor family which includes IFNγR (CDw119), IFNα/βR (CD118) and tissue factor (CD142). IL-10R has been found to play a role in signal transduction by activating specific STAT proteins (STAT3 and STAT1) and inducing gene transcription. Other reports indicate that IL-10 protein binding to NK cells appears to be stimulatory, in contrast to its binding to T cells and monocytes/macrophages. Clone 3F9 is specific for human IL-10R and its binding to the receptor can be blocked by recombinant human IL-10 (rhIL-10). This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CDw210 | |
| Monoclonal | |
| 0.5mg/mL | |
| IL-10 receptor | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| 3F9 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human IL-10R alpha Recombinant Protein | |
| 0.1 mg | |
| Cell Biology | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.