Learn More
anti-CDKN1A, SKP2 Protein Protein Interaction Antibody Pair, Abnova™
Description
Includes
CDKN1A rabbit purified polyclonal antibody (20μg); SKP2 mouse purified polyclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the CDKN1A protein, and the other against the SKP2 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
Specifications
| Antigen | CDKN1A |
| Applications | In situ PLA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Gene | CDKN1A |
| Gene Alias | CAP20/CDKN1/CIP1/MDA-6/P21/SDI1/WAF1/p21CIP1 |
| Gene Symbols | CDKN1A |
| Host Species | Mouse, Rabbit |
| Quantity | 1 Set |
| Regulatory Status | RUO |
| Vedi altri risultati |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.