missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK4 Monoclonal Antibody (DCS-35), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA512981
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MA5-12981 targets Cdk4 in IF, IP, and WB applications and shows reactivity with Human, mouse, Porcine, and Rat samples. The MA5-12981 immunogen is purified recombinant cdk4 protein.
CDK4/cyclin D3 serine/threonine kinase is dimeric protein complex. CDK4 is a member of the cyclin-dependent protein kinase family and is involved in the control of cell proliferation during the G1 phase of cell cycle. CDK4 forms a complex with the D-type cyclins and is inhibited by p16 (cyclin-dependent kinase inhibitor-2). CDK4 can mediate phosphorylation of the C-terminal region of Rb protein leading to an active transcriptional repression of E2F complex. CDC37 and HSP90 can preferentially associate with the fraction of CDK4 not bound to D-type cyclins. SMAD3 is a major physiologic substrate of the G1 cyclin-dependent kinases CDK4 and CDK2. Although CDK6 and CDK4 can both phosphorylate multiple residues in the Rb protein, they do so with different residue selectivity in vitro; CDK6 phosphorylates Thr821 while CDK4 phosphorylates Thr826 on Rb protein.
Especificaciones
| CDK4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P11802, P30285, P35426, P79432 | |
| Cdk4 | |
| Purified recombinant cdk4 protein | |
| 500 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG1 κ |
| Immunocytochemistry, Immunoprecipitation, Western Blot | |
| DCS-35 | |
| Biotin | |
| Cdk4 | |
| Cdk4; Cell division protein kinase 4; CMM3; Crk3; cyclin dependent kinase 4; cyclin-dependent kinase 4; MGC14458; p34<PSK-J3>/cdk4; PSK-J3; serine/threonine kinase; Unknown (protein for MGC:133903) | |
| Mouse | |
| Protein G | |
| RUO | |
| 100144492, 1019, 12567, 94201 | |
| 4° C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido