missing translation for 'onlineSavingsMsg'
Learn More

Cdk1 Mouse, Unlabeled, Clone: 1, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610037

 View more versions of this product

Product Code. 15875219

  • £316.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cdk1, also known as p34 [cdc2], is a ubiquitously expressed serine/threonine protein kinase. Cdk1/cdc2 has been identified as the catalytic subunit of the maturation-promoting factor (MPF), while cyclin B acts as the regulatory subunit. The binding of these two subunits is critical to the transition into M-phase of the mammalian cell cycle, and this factor's role is regulated by a series of phosphorylations and dephosphorylations. After binding to cyclin B, cdk1/cdc2 is phosphorylated on Thr-14, by Myt1, and Tyr-15, by wee1 or mik1, yielding an inactive pre-MPF complex. Phosphorylation of cdk1/cdc2 on Thr-161 is performed by a cdk7/cyclin H complex and is necessary for activation of the cdc2 complex. Dephosphorylation of Thr-14 and Tyr-15 by CDC25 occurs at the prophase/metaphase transition and completes activation of the cdc2/cyclin B complex, initiating the cell's entry into mitosis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cdk1
Monoclonal
250μg/mL
Cdc2, p34
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
1
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human p34 [cdc2] aa. 283 to 297
50 μg
Cell Biology
Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.