Learn More
CDC5 cell division cycle 5-like (S. pombe), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CDC5L.
Brand: Abnova H00000988-A01.50uL
Description
The protein encoded by this gene shares a significant similarity with Schizosaccharomyces pombe cdc5 gene product, which is a cell cycle regulator important for G2/M transition. This protein has been demonstrated to act as a positive regulator of cell cycle G2/M progression. It was also found to be an essential component of a non-snRNA spliceosome, which contains at least five additional protein factors and is required for the second catalytic step of pre-mRNA splicing. [provided by RefSeq
Sequence: ILLGGYQSRAMGLMKQLNDLWDQIEQAHLELRTFEELKKHEDSAIPRRLECLKEDVQRQQEREKELQHRYADLLLEKETLKSKFSpecifications
| CDC5 cell division cycle 5-like (S. pombe) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CDC5L. | |
| CDC5L | |
| CEF1/KIAA0432/PCDC5RP/dJ319D22.1/hCDC5 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 988 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001253 | |
| CDC5L | |
| CDC5L (NP_001244, 719 a.a. ∼ 802 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.