Learn More
CDC5L Mouse, Unlabeled, Clone: 21, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 612362
Description
Splicing, the removal of introns from pre-mRNA, is mediated by spliceosomal complexes and occurs in two distinct catalytic steps. The first step involves cleavage of the 5′ exon and the production of a lariat intermediate. In the second step, the 3′-splice site is cleaved and the exons are fused with concomitant release of the intron lariat. The spliceosome contains multiple snRNPs and a number of non-snRNP splicing factors. CDC5L is a non-snRNP component of the splicesome that is homologous to the yeast Cdc5 protein. The sequence for CDC5L contgains a helix-turn-helix DNA binding domain (DBD), four nuclear localization signals, and a hydrophilic, proline-rich central region that is similar to the transcriptional activating domain in Myb family transcription factors. CDC5L can interact with the nuclear PP1 inhibitor NIPP-1 and the WD40 domain protein PLRG1. Both of these interactions are critical for pre-mRNA splicing and may also be important for G 2/M phase transition. In addition, CDC5L translocates from the cytoplasm to the nucleus in the presence of serum. Thus, CDC5L localization and splicesome-regulating activity may be controlled by mitogen-activated signal transduction pathways.
Bioimaging, Immunofluorescence, Western Blotting
Specifications
| CDC5L | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Rat CDC5L aa. 109-303 | |
| 50 μg | |
| Cell Biology | |
| Canine, Chicken, Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| 21 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.