Learn More
anti-CDC45L, CDKN1A Protein Protein Interaction Antibody Pair, Abnova™
Description
Includes
CDC45L rabbit purified polyclonal antibody (20μg); CDKN1A mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the CDC45L protein, and the other against the CDKN1A protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
Specifications
| Antigen | CDC45L |
| Applications | In situ PLA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Gene | CDC45L |
| Gene Alias | CDC45/CDC45L2/PORC-PI-1 |
| Gene Symbols | CDC45L |
| Host Species | Mouse, Rabbit |
| Quantity | 1 Set |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.