missing translation for 'onlineSavingsMsg'
Learn More

CDC42 Mouse, Unlabeled, Clone: 44, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610929

 View more versions of this product

Product Code. 15875709

  • ONLINE EXCLUSIVE PRICE
    £311.00
    £298.29 / 1 set
    Save £12.71
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Rho family members are small GTP binding proteins that serve as molecular switches for a number of biological processes. They cycle between active GTP-bound and inactive GDP-bound states. CDC42 is a Rho family protein that was identified in membranes of human platelets and placenta. It is the homologue of CDC42Sc, which regulates initiation of bud-site assembly in Saccharomyces cerevisiae. CDC42 regulates the function of the mammalian actin cytoskeleton, allowing for efficient cytokinesis and cell morphogenesis. CDC42 and Rac1, a Ras-related GTPase, activate MEKK1, a JNK kinase kinase, which leads to the activation of several downstream components of the MAP kinase cascade leading to activation of PAK65, a CDC42- and Rac-binding protein. PAK65 interacts with CDC42/Rac1, mediates their interaction with MEKK1, and enhances MEKK1 catalytic activity. Chronic activation of CDC42 has been shown to induce malignant cellular transformation. Due to sequence homology among various small GTPase family members, potential cross-reactivity could be observed with this antibody. BD Pharmingen has not performed such cross-reactivity testing.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CDC42
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human CDC42 aa. 1-191
150 μg
Cell Biology
Canine, Human, Mouse, Rat
IgG1
Western Blot
44
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.