Learn More
CD95 (Fas/APO-1) Mouse anti-Human, APC, Clone: DX2, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 340481
Description
The CD95, clone DX2, is derived from hybridization of mouse Sp2/0 myeloma cells with spleen cells from C3H/He mice immunized with L cells transfected with CD95 antigen.
The CD95 recognizes a 48-kdalton (kDa) antigen (Fas/APO-1) that is a member of the tumor necrosis factor/nerve growth factor (TNF/NGF) superfamily. It is associated with activated cells. Cross linking of CD95 antigen on activated cells can elicit apoptosis.
Specifications
| CD95 (Fas/APO-1) | |
| Monoclonal | |
| 50μg/mL | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| Human CD95-transfected L Cells | |
| RUO | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
| Flow Cytometry | |
| DX2 | |
| APC | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.