missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAS Mouse anti-Human, Clone: UT-1, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119024
This item is not returnable.
View return policy
Description
CD95 (Fas, APO-1), a 46 kDa transmembrane glycoprotein, is a cell death receptor of the TNFR superfamily. Stimulation of CD95 results in aggregation of its intracellular death domains, formation of the death-inducing signaling complex (DISC) and activation of caspases. In type I cells caspase 3 is activated by high amounts of caspase 8 generated at the DISC, in type II cells low concentration of caspase 8 activates pathway leading to the release of cytochrome c from mitochondria and activation of caspase 3 by cytochom c. Besides its roles in induction of apoptosis, Fas also triggers pro-inflammatory cytokine responses.
Specifications
| FAS | |
| Monoclonal | |
| 1 mg/mL | |
| FAS | |
| ALPS1A, APO-1, APT1, Apo-1 Fas, CD95, FAS1, FASTM, TNFRSF6 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 355 | |
| 4° C, do not freeze | |
| Liquid |
| Flow Cytometry, Functional Assay, Western Blot | |
| UT-1 | |
| Unconjugated | |
| P25445 | |
| FAS | |
| T cell lymphoma cells HUT-78. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction