missing translation for 'onlineSavingsMsg'
Learn More

CD90.2 Rat anti-Mouse, Unlabeled, Clone: 53-2.1, BD

Product Code. 15826918
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15826918 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15826918 Supplier BD Biosciences Supplier No. 553000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

The 53-2.1 antibody reacts with the CD90.2 (Thy-1.2) alloantigen on thymocytes, most peripheral T lymphocytes, some intraepithelial T lymphocytes (IEL, DEC), epithelial cells, fibroblasts, neurons, hematopoietic stem cells, but not B lymphocytes, of most mouse strains. mAb 53-2.1 has been reported not to cross-react with Thy-1.1 (e.g., AKR/J, PL), or with rat Thy-1. CD90 is a GPI-anchored membrane glycoprotein of the Ig superfamily which is involved in signal transduction. In addition, there is evidence that CD90 mediates adhesion of
thymocytes to thymic stroma. mAb 53-2.1 has been reported to block the binding of anti-mouse CD90.2 clone 30-H12 (Cat. No. 553009) to immobilized thymocyte membranes.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD90.2
Applications Flow Cytometry
Classification Monoclonal
Clone 53-2.1
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Thy-1.2; T25; Thymus cell antigen 1, theta
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Thymus / Spleen
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.