missing translation for 'onlineSavingsMsg'
Learn More

anti-CD90.1/Thy-1.1 PerCP-Cy5.5™, Clone: HIS51, Invitrogen™

CD90.1/Thy-1.1 Monoclonal antibody A14798 from Invitrogen specifically detects CD90.1/Thy-1.1 in Mouse and Rat samples. It is validated for Flow Cytometry.

Brand:  Invitrogen A14798

Product Code. 13305644

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

CD90.1/Thy-1.1 Monoclonal antibody A14798 from Invitrogen specifically detects CD90.1/Thy-1.1 in Mouse and Rat samples. It is validated for Flow Cytometry.

CD90 (Thy 1) antigen is a GPI linked glycoprotein member of the Immunoglobulin super family. It is expressed in murine T cells, thymocytes, neural cells, Kupffer's cells and fibroblasts. Thy 1 may play a role in cell-cell or cell-ligand interactions during synaptogenesis and other events in the brain.
Host Species: Mouse
Cross Reactivity: Mouse, Rat
Isotype: IgG2a, κ
Clone: HIS51
Immunogen: rat CD90
Formulation: 25 Tests Purified Antibody (0.2mg/ml) in proprietary buffer, with 0.09% sodium azide
Synonyms: THY1, Thy-1 cell surface antigen, FLJ33325, Thy-1 T-cell antigen, Thy-1 membrane glycoprotein
Uniprot Accession: P01830, P01831
Entrez GeneID: 21838, 24832
Gene Symbol: Thy1
This product detects CD90.1/Thy-1.1 in Mouse and Rat samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is rat CD90.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD90.1/Thy-1.1
Monoclonal
PerCP-Cyanine5.5
P01830, P01831
Thy1
rat CD90
Primary
Mouse, Rat
Purified
Flow Cytometry
HIS51
25 Tests Purified Antibody (0.2mg/ml) in proprietary buffer, with 0.09% sodium azide
THY1, Thy-1 cell surface antigen, FLJ33325, Thy-1 T-cell antigen, Thy-1 membrane glycoprotein
Mouse
25 Tests
21838, 24832
Store at 4° C
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.