Learn More
CD9 Mouse anti-Human, PE, Clone: M-L13, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 341647
Description
The CD9 antibody, clone M-L13, is derived from the hybridization of mouse myeloma cells with spleen cells of mice immunized with common acute lymphoblastic leukemia (cALL) cells.
The CD9 antibody recognizes a 24-kilodalton (kDa) cell surface glycoprotein belonging to the tetraspanin family. The CD9 antigen is also known as p24, tetraspanin-29 (Tspan-29), motility-related protein-1 (MRP-1), and leucocyte antigen MIC3.
Specifications
| CD9 | |
| Monoclonal | |
| 25μg/mL | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| 50 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| M-L13 | |
| PE | |
| Mouse | |
| RUO | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.