missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD9 Monoclonal Antibody (EM-04), FITC, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA110311
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD9 Monoclonal antibody specifically detects CD9 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD9 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P40240 | |
CD9 | |
Permeabilized murine bone marrow-derived mast cells (BMMC). | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Flow Cytometry | |
EM-04 | |
FITC | |
CD9 | |
5H9; 5H9 antigen; antigen CD9; BA2; BA-2/p24 antigen; BTCC-1; CD9; CD9 antigen; CD9 antigen (p24); CD9 molecule; cell growth-inhibiting gene 2 protein; DRAP-27; GIG2; Leukocyte antigen MIC3; MIC3; motility related protein-1; Motility-related protein; MRP-1; p24; sCD 9; sCD9; soluble CD 9; soluble CD9; Tetraspanin29; Tetraspanin-29; transmembrane protein CD9; TSPAN29; Tspan-29 | |
Rat | |
Size-exclusion chromatography | |
RUO | |
12527 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |