missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD9 Monoclonal Antibody (MEM-61), Alexa Fluor™ 647, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA518154
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This antibody recognizes an epitope on second extracellular domain (EC2) of CD9 antigen.
CD9 Monoclonal antibody specifically detects CD9 in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
CD9 | |
Monoclonal | |
Alexa Fluor 647 | |
CD9 | |
5H9; 5H9 antigen; antigen CD9; BA2; BA-2/p24 antigen; BTCC-1; CD9; CD9 antigen; CD9 antigen (p24); CD9 molecule; cell growth-inhibiting gene 2 protein; DRAP-27; GIG2; Leukocyte antigen MIC3; MIC3; motility related protein-1; Motility-related protein; MRP-1; p24; sCD 9; sCD9; soluble CD 9; soluble CD9; Tetraspanin29; Tetraspanin-29; transmembrane protein CD9; TSPAN29; Tspan-29 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
928 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry, Western Blot | |
MEM-61 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P21926 | |
CD9 | |
Pre-B cell line NALM-6 | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |