Learn More
CD8b Mouse anti-Pig, PE, Clone: 295/33-25, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561484
Description
The 295/33-25 monoclonal antibody specifically binds to an epitope on the CD8α chain, a 35 kDa antigen expressed on thymocytes, peripheral T lymphocytes, and NK cells. The CD8 molecule can exist as a 70 kDa homodimer, composed of α chains, or heterodimer, composed of an α and a β chain. Two peripheral CD8+ T-cell populations can be distinguished in the pig: CD8-bright CD4-negative CTL effectors/precursors and CD8-dull CD4-positive T-helper lymphocytes. Pig NK cells express CD8 (dull staining), CD2, MHC class II, LFA-1, and asialo-GM1; but not CD3, CD4, CD5, or CD6. mAb 295/33-25 was clustered as anti-CD8b at the First International Swine CD workshop.
Specifications
| CD8b | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Pig thymocytes | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 295/33-25 | |
| PE | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Pig | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.