missing translation for 'onlineSavingsMsg'
Learn More

CD8a Mouse anti-Pig, PE, Clone: 76-2-11, BD

Product Code. 15819748
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15819748 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15819748 Supplier BD Biosciences Supplier No. 559584

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The 76-2-11 (also known as clone PT8) antibody reacts with an epitope on the CD8α chain, a 35kDa antigen expressed on thymocytes, peripheral T lymphocytes, and NK cells. The CD8 molecule can exist as a 70kDa homodimer, composed of α chains, or heterodimer, composed of an α and β chain. Cells which express the CD8αα homodimer display dimmer staining with mAb 76-2-11 than CDαβ-expressing cells. The 76-2-11 mAb does not cross-react with human or bovine cells. Two peripheral CD8+ T-cell populations can be distinguished in the pig: CD8-bright CD4- CTL effectors/precursors and CD8-dull CD4+ T-helper lymphocytes. Pig NK cells express CD8 (dull staining), CD2, MHC class II, LFA-1, and asialo-GM1, but not CD3, CD4, CD5, or CD6. mAb 76-2-11 has been reported to partially inhibit in vitro cytotoxic activity of PBL to allogeneic leukocytes, but not NK-cell-mediated lysis, and to deplete CD8+ T cells in vivo. This clone was clustered as anti-CD8a at the First International Swine CD workshop.

Host Species: Mouse
Clone: 76-2-11
Isotype: IgG2a κ
Species Reactivity: Po
Immunogen: dd miniature swine thymocytes

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8a
Applications Flow Cytometry
Classification Monoclonal
Clone 76-2-11
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen dd miniature swine thymocytes
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Pig
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.