Learn More
CD8a Mouse anti-Pig, FITC, Clone: 76-2-11, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551303
Description
The 76-2-11 (also known as clone PT8) antibody reacts with an epitope on the CD8α chain, a 35kDa antigen expressed on thymocytes, peripheral T lymphocytes, and NK cells. The CD8 molecule can exist as a 70kDa homodimer, composed of α chains, or heterodimer, composed of an α and β chain. Cells which express the CD8αα homodimer display dimmer staining with mAb 76-2-11 than CDαβ-expressing cells. The 76-2-11 mAb does not cross-react with human or bovine cells. Two peripheral CD8+ T-cell populations can be distinguished in the pig: CD8-bright CD4- CTL effectors/precursors and CD8-dull CD4+ T-helper lymphocytes. Pig NK cells express CD8 (dull staining), CD2, MHC class II, LFA-1, and asialo-GM1, but not CD3, CD4, CD5, or CD6. mAb 76-2-11 has been reported to partially inhibit in vitro cytotoxic activity of PBL to allogeneic leukocytes, but not NK-cell-mediated lysis, and to deplete CD8+ T cells in vivo. This clone was clustered as anti-CD8a at the First International Swine CD workshop.
Host Species: Mouse
Clone: 76-2-11
Isotype: IgG2a κ
Species Reactivity: Porcine
Immunogen: dd miniature swine thymocytes
Flow Cytometry
Specifications
| CD8a | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| dd miniature swine thymocytes | |
| 0.1 mg | |
| Immunology | |
| Pig | |
| IgG2a κ |
| Flow Cytometry | |
| 76-2-11 | |
| FITC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.