Learn More
CD89 Mouse anti-Human, Unlabeled, Clone: A59, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555685
Description
Reacts with the Fc receptor for IgA (FcαR), a 55-75kDa glycoprotein expressed exclusively on cells of granulocytic and monocyte/macrophage lineages in peripheral blood, but not on lymphocytes. It is also expressed on promyelocytes in bone marrow and in the cytoplasm of fixed monocytes. This mAb does not block IgA binding and is more efficient than native IgA ligands in the isolation of FcαR molecules. FcαR plays a role in triggering phagocytosis and respiratory burst. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: A59
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry, Immunohistochemistry (Frozen)
Specifications
| CD89 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| A59 | |
| Unconjugated | |
| Mouse | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.