Learn More
CD88 Mouse anti-Human, Unlabeled, Clone: D53-1473, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 550493
Description
Reacts with a 40kDa polypeptide expressed on granulocytes, monocytes and macrophages. CD88 is a G type protein-coupled receptor with seven transmembrane domains. It binds to C5a, the α chain of the complement component C5. C5a is cleaved from C5 during complement activation. The binding of C5a to C5a receptor (C5aR, CD88) on granulocytes leads to various effects including shedding of selectins, upregulation of adhesion molecules, chemotaxis, granule exocytosis and activation of NADPH oxidase. Clone D53-1473 was developed using a known C5aR N-terminal extracellular hydrophilic region (a.a. 1-31) peptide as immunogen. This antibody is useful in inflammation and chemotaxis research. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: D53-1473
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry, Immunofluorescence
Specifications
| CD88 | |
| Monoclonal | |
| 0.5mg/mL | |
| C5a Receptor | |
| Mouse | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| D53-1473 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.