missing translation for 'onlineSavingsMsg'
Learn More

CD86 Mouse anti-Human, V450, Clone: 2331 (FUN-1), BD

Mouse Monoclonal Antibody

£183.61 - £318.26

Specifications

Antigen CD86
Clone 2331 (FUN-1)
Applications Flow Cytometry
Classification Monoclonal
Conjugate Violet Fluor 450 (V450)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15800139
View Documents
BD Biosciences
560359
30 Tests
ONLINE EXCLUSIVE PRICE
£192.00
£183.61 / Each
Save £8.39
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15880129
View Documents
BD Biosciences
560357
120 Tests
ONLINE EXCLUSIVE PRICE
£331.00
£318.26 / Each
Save £12.74
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2331 (FUN-1) recognizes a 75kDa cell surface protein, CD86 (B70/B7-2), expressed primarily on monocytes and activated B cells. Competitive binding assays demonstrate that, while both 2331 (FUN-1) and IT2.2 (anti-CD86, Mfr. No. 555663) recognize the same molecule, they react with different epitopes. CD86 is the second ligand for CD28 and CTLA-4 and may play an important role in co-stimulation of T cells in primary immune response. 2331 (FUN-1) blocks costimulation activity of CD86 in functional studies. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.

Host Species: Mouse
Clone: 2331(FUN-1)
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

Specifications

Specifications

CD86
Flow Cytometry
Violet Fluor 450 (V450)
RUO
Human
Human HBL-1 Cell Line
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
2331 (FUN-1)
Monoclonal
Mouse
Immunology
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
B7.2; B7-2; B-lymphocyte activation antigen B7-2; B70; BU63; CD28LG2; LAB72
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.