Learn More
CD86 Mouse anti-Human, Brilliant Violet 421, Clone: 2331 (FUN-1), BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562433
Description
The 2331 (FUN-1) monoclonal antibody specifically recognizes a 75 kDa transmembrane cell surface protein, CD86 (B70/B7-2), expressed primarily on monocytes, dendritic cells and activated B cells. Competitive binding assays demonstrate that, while both 2331 (FUN-1) and IT2.2 (anti-CD86, Cat. No. 555663) antibodies specifically recognize the same molecule, they react with different epitopes. CD86 is the second ligand for CD28 and CTLA-4 and may play an important role in co-stimulation of T cells in primary immune response. The 2331 (FUN-1) antibody blocks the costimulatory activity of CD86 when tested in functional studies.The antibody was conjugated to Brilliant Violet 421 and has been developed in collaboration with Sirigen. With an Ex Max of 407-nm and Em Max at 421-nm, Brilliant Violet 421 can be excited by the violet laser and detected in the standard Pacific Blue filter set (eg, 450/50-nm filter). Brilliant Violet 421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.
Host Species: Mouse
Clone: 2331 (FUN-1)
Isotype: IgG1, κ
Species Reactivity: Human
Flow Cytometry
Specifications
| CD86 | |
| Monoclonal | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human HBL-1 Cell Line | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 2331 (FUN-1) | |
| B7.2; B7-2; B-lymphocyte activation antigen B7-2; B70; BU63; CD28LG2; LAB72 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.