missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD86; Clone: 2331 (FUN-1); BD
Brand: BD Biosciences 562999
This item is not returnable.
View return policy
Description
Host Species: Mouse
Clone 2331 (FUN-1)
Isotype IgG1, κ
Species Reactivity Hu
Immunogen Human HBL-1 Cell Line
Intracellular block/Flow Cytometry
Specifications
| CD86 | |
| 2331 (FUN-1) | |
| Human HBL-1 Cell Line | |
| 100 Tests | |
| Primary | |
| IgG1 |
| Monoclonal | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction