missing translation for 'onlineSavingsMsg'
Learn More

CD81 Hamster anti-Mouse/Rat, PE, Clone: EAT2, BD

Hamster Monoclonal Antibody

Brand:  BD Biosciences 559519

Product Code. 15809738

  • ONLINE EXCLUSIVE PRICE
    £467.00
    £449.04 / Each
    Save £17.96
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The Eat2 antibody reacts with CD81, a 26kDa nonglycosylated member of the transmembrane 4 integral membrane protein superfamily, expressed by many types of cells. For example, CD81 participates with CD19 and CD21 in the signal transduction complex associated with the B-cell receptor on human B lymphocytes and with the CD4 and CD8 co-receptors on human thymocytes and T lymphocytes. In mouse fetal thymic organ culture, interactions of immature thymocytes with CD81 expressed by thymic stromal cells are required to induce development of T cells with αβ T-cell receptors. Furthermore, CD81 has been shown to play a role in the regulation of rat mastcell degranulation. Despite its important roles in the immune response and wide tissue distribution, CD81-deficient mice are born without obvious developmental abnormalities. However, these mice have abnormal immune responses, and impaired fertility. Eat2 mAb cross-reacts with the rat CD81 antigen. Eat2 mAb was reported cross-reacting with the rat CD81 antigen.

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD81
Monoclonal
0.2mg/mL
TAPA-1; Tspan28; Tapa1; Tapa-1; Target of the antiproliferative antibody 1;
Hamster
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
EAT2
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
CD81+ mouse B lymphoma 38C13
0.1 mg
Immunology
Mouse, Rat
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.