Learn More
CD80 Mouse anti-Rat, PE, Clone: 3H5, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555014
Description
The 3H5 antibody reacts with CD80 (B7-1), a member of the Ig superfamily of transmembrane proteins. CD80, a ligand for CD28 and CD152 (CTLA-4), is one of the accessory molecules that plays an important role in T cell-B cell costimulatory interactions. CD80 is predominantly expressed on antigen-presenting cells. It can be induced on splenic B cells by in vitro polyclonal activation such as LPS treatment. 3H5 mAb is reported to block the costimulatory function of rat CD80 and to immunoprecipitate CD80 from Lewis-S1 cell lysates.
Flow Cytometry
Specifications
| CD80 | |
| Monoclonal | |
| 0.2mg/mL | |
| B7-1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 3H5 | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat HTLV-1-transformed T-cell line Lewis-S1 | |
| 0.2 mg | |
| Immunology | |
| Rat | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.