Learn More
CD80 Hamster anti-Mouse, APC, Clone: 16-10A1, BD
Hamster Monoclonal Antibody
Brand: BD Biosciences 560016
Beskrivning
The 16-10A1 antibody reacts with CD80 (B7-1). This member of the Ig superfamily, along with CD86 (B7-2), participates in T-cell co-stimulation via interactions with CD28 and CD152 (CTLA-4). CD80 has been reported to be constitutively expressed on dendritic cells, monocytes, and peritoneal macrophages; and it is inducible on B cells by various means, including activation by LPS, IL-4, and the cross-linking of surface Ig. Expression of CD80 has been reported to be greatly enhanced on splenic B cells following activation by LPS, with peak expression occurring between 48 and 72 hours. It has been reported that the activation of purified B cells with LPS can induce CD80 expression in as few as 18 hours. The 16-10A1 antibody has been reported to block binding of CTLA-4 Ig to CD80 and to block T-cell activation by Con A-elicited peritoneal exudate cells and CD80-transfected cell lines. However, 16-10A1 antibody alone is not able to block T-cell activation by antigen-presenting cells. CD86 (B7-2) is an alternate ligand for CD28 and CD152 (CTLA-4). Preliminary reports indicate that the 16-10A1 mAb may block the binding of rat anti-CD80 mAb clone 1G10 (Cat. No. 553368). In addition, it has been reported that the 16-10A1 antibody may cross-react with an activation antigen expressed on IFN-γ-activated alveolar macrophages of the dog.
Specifikationer
| CD80 | |
| Monoclonal | |
| 0.2mg/mL | |
| Cd80; B7; B7-1; Cd28l; Ly-53; MIC17; TSA1 | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Canine, Mouse | |
| IgG2 κ |
| Flow Cytometry | |
| 16-10A1 | |
| APC | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse CD80 (B7) Transfected Cell Line | |
| 0.1 mg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.