Learn More
CD8 Mouse anti-Human, Unlabeled, Clone: RPA-T8, BD
Mouse Monoclonal Antibody
£106.21 - £162.90
Specifications
| Antigen | CD8 |
|---|---|
| Clone | RPA-T8 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15838838
|
BD Biosciences
557084 |
0.1 mg |
£170.00 £162.90 / Each Save £7.10 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15888098
|
BD Biosciences
555364 |
0.5 mg |
£179.00 £106.21 / Each Save £72.79 40% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Clone RPA-T8 reacts with the ? subunit (32kDa) of the two-chain complex expressed on T-cytoxic/suppressor cell populations. CD8 molecule binds to HLA class I molecules during interaction of CD8+ T cells with antigen-presenting cells or with target cells. RPA-T8 reacts with approximately 13-48% of peripheral blood lymphocytes and 80% of thymocytes, as well as a subset of NK cells. RPA-T8 and HIT8a (Cat. No. 555337) are not cross-blocking.
Host Species: Mouse
Clone: RPA-T8
Isotype: IgG1, κ
Cross Reactivity: Baboon, Cynomolgus
Flow Cytometry
Specifications
| CD8 | |
| 0.5mg/mL | |
| Monoclonal | |
| Mouse | |
| Immunology | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| CD8α; CD8A; CD8 alpha; Leu2; MAL; T8; p32 |
| RPA-T8 | |
| Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Baboon, Cynomolgus, Human, Rhesus Monkey | |
| Human CD8a | |
| Primary | |
| Store undiluted at 4°C |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.