missing translation for 'onlineSavingsMsg'
Learn More

CD8, BUV737, Mouse anti-Human, Clone: SK1, BD

Product Code. 15883759
Change view
Click to view available options
Quantity:
25 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15883759 25 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15883759 Supplier BD Biosciences Supplier No. 564628

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Applications:

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8
Applications Flow Cytometry
Classification Monoclonal
Clone SK1
Conjugate Brilliant Ultraviolet 737
Host Species Mouse
Immunogen Human Peripheral Blood T Cells
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4deg.C and protected from prolonged exposure to light. Do not freeze
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.