missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD8, Alexa Fluor 750, Clone: 53-6.7, R&D Systems™
Rat Monoclonal Antibody
Specifications
| Antigen | CD8 |
|---|---|
| Immunogen | Mouse thymus or spleen |
| Clone | 53-6.7 |
| Applications | Flow Cytometry |
| Conjugate | Alexa Fluor 750 |
Specifications
| CD8 | |
| 53-6.7 | |
| Alexa Fluor 750 | |
| Rat | |
| Primary | |
| Detects both the alpha and alpha ' chains of mouse CD8 (1). | |
| Protein A or G purified from hybridoma culture supernatant |
| Mouse thymus or spleen | |
| Flow Cytometry | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| IgG2a | |
| 925 | |
| RUO |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title