Learn More
CD79a Mouse anti-Human, Unlabeled, Clone: HM47, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555934
Description
The HM47 monoclonal antibody specifically binds to the cytoplasmic domain of CD79a. CD79a is a 47 kDa type 1 transmembrane glycoprotein present on B lymphocytes. CD79a is also referred to as mb-1, IGA and Ig-alpha (Ig α ). It is expressed on B cells at various stages of differentiation, from pre-B cell stage, probably before expression of cytoplasmic µ chain, to plasma cell stage, in which it is detected only in the cytoplasm. CD79a associates with CD79b to form part of the B-cell receptor complex. It has been suggested that CD79a may play a role in mediating the transport of IgM to the cell surface. This antibody has been found to crossreact with permeabilized A20 cells (mouse B cell line).
Specifications
| CD79a | |
| Monoclonal | |
| 0.5mg/mL | |
| CD79a, immunoglobulin-associated alpha ; CD79A; Ig-alpha; IGA; Igα; MB-1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| HM47 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human CD79a Peptide | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.