missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST6GAL1 Mouse anti-Human, Clone: LN-1, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA135289
Code : LD
Additional Details : Weight : 0.01000kg
Description
A suggested positive control for this product is human tonsil.
Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.Specifications
ST6GAL1 | |
Monoclonal | |
0.1 mg/mL | |
ST6GAL1 | |
Sialyltransferase 1, SIAT1, ST6GalI, ST6N, B-cell antigen CD75, CMP-N-acetylneuraminate beta-galactosamide alpha-2, 6-sialyltransferase, CMP-N-acetylneuraminate-beta-galactosamide-alpha-2, 6-sialyltransferase 1, ST6Gal I, alpha 2, 6-ST 1, beta-galactoside alpha-2, 6-sialyltransferase 1, sialyltransferase 1 | |
Mouse | |
Purified | |
RUO | |
6480 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
LN-1 | |
Unconjugated | |
P15907 | |
ST6GAL1 | |
Lymphocytes/SU-DHL4 cell line. | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgM |