Learn More
CD74 Mouse anti-Human, Unlabeled, Clone: M-B741, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555538
Description
Reacts with cell -surface MHC class II-associated invariant chain molecule (41, 35, 33 kD). CD74 is moderately expressed on B cells, weakly on monocytes and macrophages. Other reactive cells include Langerhans cells, dendritic cells, a subset of activated T cells and thymic epithelium. CD74 is postulated to play a role in antigen processing and presentation through intracellular transport and expression of class II molecules on the cell surface.
Specifications
| CD74 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| M-B741 | |
| Unconjugated | |
| Mouse | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.