Learn More
CD72 Mouse anti-Human, FITC, Clone: J4-117, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555918
Description
Reacts with a 39-43 kDa type II integral membrane protein expressed on B cells from early precursor stages through mature B cells. CD72 is not expressed on plasma cells. CD72 has been reported to bind to CD5, a glycoprotein expressed on all mature T cells and a subpopulation of B cells, suggesting a role in regulating T- and B-cell activation and proliferation
Specifications
| CD72 | |
| Monoclonal | |
| FITC | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG2b κ |
| Flow Cytometry | |
| J4-117 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.