Learn More
CD71 (Transferrin Receptor) Mouse anti-Human, Unlabeled, Clone: L01.1, BD
Mouse Monoclonal Antibody
Brand:  BD Biosciences 347510
Product Code. 15896098
  • £621.00 / Each
Description
The CD71 antibody, clone L01.1, is derived from the hybridization of P3-X63- Ag8.653 mouse myeloma cells with spleen cells from BALB/c mice immunized with blast cells from pokeweed mitogen–stimulated peripheral blood cells.
The CD71 antibody is specific for the human transferrin receptor, with a molecular weight of 190 kilodaltons (kDa), 90-kDa reduced.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD71 (Transferrin Receptor)
Monoclonal
100μg/mL
Transferrin Receptor
Mouse
Affinity Purified
RUO
Primary
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Flow Cytometry
L01.1
Unconjugated
Phosphate buffered saline with gelatin and 0.1% sodium azide.
Blast cells from pokeweed mitogen-stimulated peripheral blood cells
100 Tests
Immunology
Human
IgG2a κ