Learn More
CD69 Mouse anti-Human, APC, Clone: L78, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 340560
Description
CD69, clone L78, is derived from hybridization of mouse Sp2/0-Ag14 myeloma cells with lymph node cells from BALB/c mice immunized with a CD8+ alloantigen–directed cytotoxic T-lymphocyte (CTL) cell line.
CD69 recognizes a very early human activation antigen that is a disulfide-bonded homodimer consisting of Mr 60-kilodalton (kd) polypeptides with one or two N-linked oligosaccharides.
Specifications
| CD69 | |
| Monoclonal | |
| 200μg/mL | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| CD8 | |
| RUO | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
| Flow Cytometry | |
| L78 | |
| APC | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.